EMAIL THIS PAGE TO A FRIEND
Journal of materials science. Materials in medicine
Carbonate-containing hydroxyapatite derived from calcium tripolyphosphate gel with urea.
PMID 15965739
Abstract
Carbonate containing hydroxyapatite (CO3HAp) is one of the candidate materials as a bioresorbable bone substitute. In the present work, CO3HAp was efficiently prepared by a hydrothermal treatment of calcium tripolyphosphate gel with urea at 140 degrees C for 24 h. Chemical potential plots of the CO3HAp for estimation of its dissolution behavior suggested that the CO3HAp is more soluble than hydroxyapatite (HAp) and is as soluble as octacalcium phosphate (OCP) and/or beta -tricalcium phosphate (TCP). This material is expected to be applied to bioresorbable materials such as bone fillers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T5508
Trisodium trimetaphosphate, ≥95%
Na3O9P3