EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Increased levels of inositol hexakisphosphate (InsP6) protect HEK293 cells from tumor necrosis factor (alpha)- and Fas-induced apoptosis.
PMID 15967797
Abstract
The overexpression of inositol 1,3,4-trisphosphate 5/6-kinase has recently been shown to protect HEK293 cells from tumor necrosis factor alpha (TNF(alpha))-induced apoptosis. This overexpression leads to an increase in the levels of both inositol 1,3,4,5,6-pentakisphosphate (InsP5) and inositol 1,2,3,4,5,6-hexakisphosphate (InsP6). Cells that overexpress InsP5 2-kinase have increased levels of InsP6 and are also protected from TNFalpha-induced apoptosis; furthermore, cells that express an RNA interference construct to the 2-kinase are deficient in InsP6 and are sensitized to TNFalpha-induced apoptosis. Therefore the protective effect of 5/6-kinase on TNFalpha-mediated apoptosis is due to an increase of InsP6 or to a metabolite derived from InsP6. Furthermore, we find that the InsP6 also protects from Fas-mediated apoptosis. No effect was seen in the endocytic rate of transferrin receptor, caspase 8 activity, or TNF receptor number at the cell surface. Cells that overexpress 2-kinase do show an increase in the amount of receptor-interacting protein (RIP), while cells with reduced InsP6 levels show relatively less RIP, providing a possible mechanism for the effect on apoptosis.