EMAIL THIS PAGE TO A FRIEND
Pharmacogenetics and genomics
Theophylline pharmacokinetics: comparison of Cyp1a1(-/-) and Cyp1a2(-/-) knockout mice, humanized hCYP1A1_1A2 knock-in mice lacking either the mouse Cyp1a1 or Cyp1a2 gene, and Cyp1(+/+) wild-type mice.
PMID 15970798
Abstract
Pharmacokinetics of theophylline was investigated in Cyp1(+/+) wild-type mice, Cyp1a1(-/-) and Cyp1a2(-/-) knockout mice, and humanized hCYP1A1_1A2 mice lacking either the mouse Cyp1a1 or Cyp1a2 gene. Animals received a single dose of theophylline (8 mg/kg i.p.), either alone or pretreated with 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD; 10 microg/kg i.p.) 24 h prior to theophylline. We found that mouse or human CYP1A2 is the predominant enzyme for theophylline metabolism, the contribution of mouse or human CYP1A1 to theophylline metabolism is negligible, and another TCDD-inducible enzyme plays a minor role in 1-methyluric acid and 1,3-dimethyluric acid formation as well as enhanced theophylline clearance from the body. The half-life of elimination from plasma was more than four times longer in Cyp1a2(-/-) than Cyp1(+/+) mice and more than 10 times different after TCDD pretreatment. In humanized hCYP1A1_1A2 mice lacking the mouse Cyp1a2 gene, the half-life of elimination from plasma was two to three times longer than that in Cyp1(+/+) mice and four to five times different after TCDD pretreatment. Replacement of mouse Cyp1a2 with a functional human CYP1A2 gene restored the ability to metabolize theophylline, and the metabolism changed to a humanized profile (i.e. 3-methylxanthine formation, not seen in the wild-type mouse). TCDD-pretreated hCYP1A1_1A2 Cyp1a2(-/-) mice exhibited enhanced theophylline metabolism and clearance, due to induction of the human CYP1A2 enzyme. Comparing the hCYP1A1_1A2 Cyp1a2(-/-) and wild-type mice with published clinical studies, we found theophylline clearance to be about 5 times and 12 times, respectively, greater than that reported in humans.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
222526
3-Methylxanthine, 98%
C6H6N4O2