EMAIL THIS PAGE TO A FRIEND
American journal of obstetrics and gynecology
Effect of oral versus transdermal steroidal contraceptives on androgenic markers.
PMID 15970897
Abstract
The purpose of this study was to compare biochemical androgen profiles in women treated with the contraceptive patch versus an oral contraceptive (OC). Twenty-four healthy women were randomly assigned to receive 3 cycles of either the contraceptive patch (ethinyl estradiol [EE] 20 microg/d and norelgestromin 150 microg/d) or OC (EE 35 mug and norgestimate 250 microg). Blood samples were taken at baseline and end of treatment. Serum levels of sex hormone-binding globulin (SHBG), total testosterone (T), androstenedione (A), dehydroepiandrosterone sulfate (DHEAS), dihydrotestosterone (DHT), and 3alpha-androstanediol glucuronide (3alpha-diol G) were quantified by immunoassay methods; free T was calculated. The paired t and Student t tests were used for statistical analysis. Nineteen women completed the study (patch, n = 10; OC, n = 9). Despite a 1.6-fold relative increase in SHBG levels with the patch versus OC (449% vs 274%, P = .03), free T decreased equally in both groups (patch 60%, P < .0001; OC 59%, P < .0001). DHEAS decreased by 26% in the patch group (P < .01) and 32% in the OC group (P < .001). 3alpha-diol G was reduced by 52% in the patch group (P < .0001) and 51% in the OC group (P < .0001). In addition, the OC was associated with significant decreases in A and DHT. The contraceptive patch had an effect comparable to the OC on several key androgenic markers. Given these biochemical findings, the contraceptive patch has significant potential as a therapeutic agent for disorders of androgen excess.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E1001
Ethisterone, ≥99% (HPLC)
C21H28O2