EMAIL THIS PAGE TO A FRIEND
Journal of industrial microbiology & biotechnology
Dextran dextrinase and dextran of Gluconobacter oxydans.
PMID 15973532
Abstract
Certain strains of Gluconobacter oxydans have been known since the 1940s to produce the enzyme dextran dextrinase (DDase; EC2.4.1.2)-a transglucosidase converting maltodextrins into (oligo)dextran. The enzyme catalyses the transfer of an alpha1,4 linked glucosyl unit from a donor to an acceptor molecule, forming an alpha1,6 linkage: consecutive glucosyl transfers result in the formation of high molecular weight dextran from maltodextrins. In the early 1990s, the group of K. Yamamoto in Japan revived research on DDase, focussing on the purification and characterisation of the intracellular DDase produced by G. oxydans ATCC 11894. More recently, this was taken further by Y. Suzuki and coworkers, who investigated the properties and kinetics of the extracellular DDase formed by the same strain. Our group further elaborated on fermentation processes to optimise DDase production and dextran formation, DDase characterisation and its use as a biocatalyst, and the physiological link between intracellular and extracellular DDase. Here, we present a condensed overview of the current scientific status and the application potential of G. oxydans DDase and its products, (oligo)dextrans. The production of DDase as well as of dextran is first described via optimised fermentation processes. Specific assays for measuring DDase activity are also outlined. The general characteristics, substrate specificity, and mode of action of DDase as a transglucosidase are described in detail. Two forms of DDase are produced by G. oxydans depending on nutritional fermentation conditions: an intracellular and an extracellular form. The relationship between the two enzyme forms is also discussed. Furthermore, applications of DDase, e.g. production of (oligo)dextran, transglucosylated products and speciality oligosaccharides, are summarized.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31430 Dextran, analytical standard, for GPC, Set Mp 1,000-400,000
00268 Dextran, analytical standard, for GPC, 1,000
00891 Dextran, analytical standard, for GPC, 50,000
00269 Dextran, analytical standard, for GPC, 5,000
00270 Dextran, analytical standard, for GPC, 12,000
00894 Dextran, analytical standard, for GPC, 270,000
00271 Dextran, analytical standard, for GPC, 25,000
00895 Dextran, analytical standard, for GPC, 410,000
00892 Dextran, analytical standard, for GPC, 80,000
00893 Dextran, analytical standard, for GPC, 150,000
00896 Dextran, analytical standard, for GPC, 670,000
31394 Dextran, enzymatic synth.
D0731000 Dextran, European Pharmacopoeia (EP) Reference Standard
D0731005 Dextran 1, European Pharmacopoeia (EP) Reference Standard
D0734000 Dextran 10 for calibration, European Pharmacopoeia (EP) Reference Standard
D0737000 Dextran 250 for calibration, European Pharmacopoeia (EP) Reference Standard
D0733000 Dextran 4 for calibration, European Pharmacopoeia (EP) Reference Standard
D0735000 Dextran 40 for calibration, European Pharmacopoeia (EP) Reference Standard
D0738000 Dextran 40 for performance test, European Pharmacopoeia (EP) Reference Standard
D0739000 Dextran 60/70 for performance test, European Pharmacopoeia (EP) Reference Standard
D0736000 Dextran 70 for calibration, European Pharmacopoeia (EP) Reference Standard
D8821 Dextran from Leuconostoc mesenteroides, average mol wt 60,000-76,000
31398 Dextran from Leuconostoc mesenteroides, Mr ~200,000
31416 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 1,000
31417 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 5,000
31397 Dextran from Leuconostoc mesenteroides, Mr ~60,000
31419 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 25,000
31420 Dextran from Leuconostoc mesenteroides, for GPC, analytical standard, Mw 50,000
31425 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 670,000
31418 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 12,000