EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
Purification and characterization of a novel enoyl coenzyme A reductase from Streptomyces collinus.
PMID 1597409
Abstract
A novel NADPH-dependent enoyl reductase, catalyzing the conversion of 1-cyclohexenylcarbonyl coenzyme A (1-cyclohexenylcarbonyl-CoA) to cyclohexylcarbonyl-CoA, was purified to homogeneity from Streptomyces collinus. This enzyme, a dimer with subunits of identical M(r) (36,000), exhibits a Km of 1.5 +/- 0.3 microM for NADPH and 25 +/- 3 microM for 1-cyclohexenylcarbonyl-CoA. It has a pH optimum of 7.5, is most active at 30 degrees C, and is inhibited by both divalent cations and thiol reagents. Two internal peptide sequences were obtained. Ansatrienin A (an antibiotic produced by S. collinus) contains a cyclohexanecarboxylic acid moiety, and it is suggested that the 1-cyclohexenylcarbonyl-CoA reductase described herein catalyzes the final reductive step in the conversion of shikimic acid into this moiety.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
328367
1-Cyclohexene-1-carboxylic acid, 97%
C7H10O2