EMAIL THIS PAGE TO A FRIEND
The New England journal of medicine
Hypothyroidism due to ethionamide.
PMID 15987931
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E6005
Ethionamide
C8H10N2S