EMAIL THIS PAGE TO A FRIEND
Journal of peptide science : an official publication of the European Peptide Society
Kinetic investigation of the effect of the amino acid side chains in the selective acidolysis of N-acyl-N,alpha,alpha-trialkyl glycine amides.
PMID 15988788
Abstract
Accurate kinetic measurements of the rate constants for the acidolysis of five N-acetyl-N-(4-methoxybenzyl)-alpha,alpha-trialkyl glycine cyclohexyl amides in TFA were performed at 25 degrees C and the reactions monitored by HPLC. In all cases the results were consistent with a first order behaviour with respect to the substrate. No direct correlation was obtained with these data between the rate constant values and structure, but a good correlation coefficient was obtained when a multiple regression analysis was applied by taking advantage of a Taft equation using appropriate polar and steric substituent parameters. In a plot of the values observed for log k against those calculated by this equation all five points fell very close to the line of perfect correlation. The calculated sensitivity coefficients to polar and steric contributions were used to discuss the experimental results and showed that the acidolysis were comparatively less affected by steric effects than expected.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50070
Glycinamide hydrochloride, ≥99.0% (AT)
C2H6N2O · HCl
G6104
Glycinamide hydrochloride, 98%
C2H6N2O · HCl