EMAIL THIS PAGE TO A FRIEND
Journal of the American Veterinary Medical Association
Questions limitation in treatment of hepatozoonosis.
PMID 15989174
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33823
Decoquinate, VETRANAL, analytical standard
C24H35NO5