EMAIL THIS PAGE TO A FRIEND
Chemosphere
Inhibition of laccase activity from Trametes versicolor by heavy metals and organic compounds.
PMID 15993161
Abstract
Due to the numerous biotechnological applications of laccase enzyme, it is essential to know the influence of different agents usually present in the natural environment on its enzymatic action, especially for in situ treatment technologies. In the present work, a simple and rapid method to determine the inhibitory or inducer effect of different compounds on laccase activity was developed. The compounds tested were copper-chelating agents and heavy metals. It was found that using syringaldazine as a substrate, all copper-chelating agents (except EDTA) highly inhibited laccase activity (around 100%) at an inhibitor concentration lower than 20 mM. Moreover, 40% of inhibition, which was detected at a concentration of 20 mM for both Cd(2+) and Cu(2+) increased with concentration until nearly complete inhibition at 80 mM.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S7896
Syringaldazine, indicator for laccase and peroxidase activity
C18H20N2O6
177539
Syringaldazine, 98%
C18H20N2O6