EMAIL THIS PAGE TO A FRIEND
Chemosphere
Direct Yellow and Methylene Blue liquid-liquid extraction with alkylene carbonates.
PMID 15993164
Abstract
Propylene and 1,2-butylene carbonates enable very effective extraction of Methylene Blue. The use of a more hydrophobic 1,2-butylene carbonate is preferred for practical application. Separation is enhanced by addition of an electrolyte (NaCl). Partition coefficients for solutions containing Methylene Blue and 1M NaCl are at least a hundred times higher than for Direct Yellow solutions. The recovery of Methylene Blue with propylene and 1,2-butylene carbonates is significantly better, compared with the cloud point technique. Cloud point technique yields to almost 100% only in the case of low concentrations of Methylene Blue in the feed (0.5mg dm(-3)). For higher initial concentrations (0.01-0.25 g dm(-3)) of Methylene Blue, this method of recovery provides low extraction percentages (10-40%). Alkylene carbonates are not appropriate extractants for recovery of very hydrophilic Direct Yellow.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
517135
Diethyl carbonate, anhydrous, ≥99%
C5H10O3
D91551
Diethyl carbonate, 99%
C5H10O3