EMAIL THIS PAGE TO A FRIEND
Toxicology and applied pharmacology
Current and emerging challenges in toxicopathology: carcinogenic threshold of phenobarbital and proof of arsenic carcinogenicity using rat medium-term bioassays for carcinogens.
PMID 15993454
Abstract
For the last 25 years, Prof. Nobuyuki Ito and his laboratory have focused on the development of liver medium-term bioassay system for detection of carcinogens in F344 rats utilizing glutathione S-transferase placental form (GST-P)-positive foci as an end point marker. In this presentation, the outline and samples of medium-term bioassay systems were described. Furthermore, our data demonstrated the presence of a threshold for the non-genotoxic carcinogen, phenobarbital (PB), and the lack of linearity in the low-dose area of the dose-response curve, providing evidence for hormesis. In addition, the establishment and applications of multiorgan carcinogenicity bioassay (DMBDD model), used for the examination of the carcinogenicity of genotoxic and non-genotoxic chemicals, are discussed. Dimethylarsinic acid, one of organic arsenics, was found to be carcinogenic in rat bladder using DMBDD model and carcinogenicity test.