EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Synthesis and spectroscopic study of copper(II) and manganese(II) complexes with pipemidic acid.
PMID 15996512
Abstract
The interaction of copper(II) and manganese(II) with pipemidic acid, Hpipem, afforded the complexes [Cu(pipem)(2)(H(2)O)] x 2H(2)O, 1 and [Mn(pipem)(2)(H(2)O)], 2. The new complexes have been characterised by elemental analyses, infrared, UV-vis and X-band EPR spectroscopy in the temperature range from 4 to 300 K. The monoanion, pipem, exhibits O,O ligation through the carbonyl and carboxylato oxygen atoms. Five coordinate square-pyramid configuration has been proposed for 1 and 2, and the fifth apical position is occupied by a coordinated water molecule.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P7903
Pipemidic acid
C14H17N5O3