EMAIL THIS PAGE TO A FRIEND
Chemosphere
Monitoring tricyclazole residues in rice paddy watersheds.
PMID 15996714
Abstract
Pesticide application to rice paddies may affect the quality of environmental resources such as groundwater and surface water. The distribution of residues of tricyclazole, an environmentally persistent fungicide used widely in Italy, was monitored in the network of surface water bodies surrounding the main rice production area in Italy. The location of monitoring sites was based on the potential risk for contamination with tricyclazole. This was determined as a function of the area of rice grown, the geographical distribution of rice crops susceptible to the pest, and sales of tricyclazole. Monitoring sites were also located to represent different spatial scales (farm, catchment and basin). For water samples taken shortly after application in July and August, the highest concentrations of tricyclazole were measured at the farm sites. However, residues were also detected at the catchment and basin scale. The 95% of the measured residue levels was below 9.80, 1.20 and 1.15 microg l(-1), at the farm, catchment and basin scales, respectively. In sediment, tricyclazole residues were detected in 12 out 176 samples collected with the 95% of the measured residue levels below the concentration of 0.03 mg kg(-1). Residues were sporadically detected in samples taken after the crop was harvested in November and December. Variables such as the scale of sampling, the season and the year, were significant in determining pesticide residue distribution. The type of water body was less significant.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45808
Tricyclazole, PESTANAL®, analytical standard
C9H7N3S