EMAIL THIS PAGE TO A FRIEND
Neuropharmacology
Analgesic effects of the selective group II (mGlu2/3) metabotropic glutamate receptor agonists LY379268 and LY389795 in persistent and inflammatory pain models after acute and repeated dosing
PMID 15998527