EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Solid-phase synthesis and anti-infective activity of a combinatorial library based on the natural product anisomycin.
PMID 16005213
Abstract
The solid-phase synthesis of a library based on the natural product anisomycin is described. The resulting library was tested against a panel of bacterial and fungal targets, and active compounds were identified in a Staphylococcus aureus whole-cell assay and an efflux-deficient fungal whole-cell assay.