EMAIL THIS PAGE TO A FRIEND
Guang pu xue yu guang pu fen xi = Guang pu
[Research on energy transfer between riboflavin and anthrone derivative].
PMID 16013301
Abstract
According to Forster's theory about energy transfer, the energy transfer between riboflavin and anthrone derivative was investigated. It was found that there is a notable energy transfer between riboflavin and anthrone derivative, and the energy transfer efficiency is affected by the following factors: heating temperature, heating time, and mass ratio. And the best ratio of riboflavin and anthrone derivative is 1:3. At the same time the energy transfer efficiency drops with increasing heating temperature and time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
319899
Anthrone, ACS reagent, 97%
C14H10O