EMAIL THIS PAGE TO A FRIEND
Indian heart journal
Anti-arrhythmic effects of herbal medicine.
PMID 16013349
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T9517
Glyceryl trilinoleate, ≥98% (TLC), liquid
C57H98O6