EMAIL THIS PAGE TO A FRIEND
Se pu = Chinese journal of chromatography
[Determination of bromodichloromethane and dibromochloromethane in tap water using headspace gas chromatography with standard addition method].
PMID 16013579