EMAIL THIS PAGE TO A FRIEND
Expert opinion on pharmacotherapy
Miltefosine to treat leishmaniasis.
PMID 16013987
Abstract
Leishmaniasis exists in both visceral and cutaneous forms, and miltefosine is the first oral agent with demonstrable efficacy against both types of this disease. At a dose of approximately 2.5 mg/kg/day for 28 days, miltefosine is > 90% curative for visceral disease in India and cutaneous disease in Colombia. Miltefosine is a lecithin analogue and its mechanism may be to inhibit phosphatidylcholine biosynthesis in the causative parasites. The clinical half-life of miltefosine is approximately 7 days. Whether or not miltefosine can be used for widespread out-patient treatment of individuals and whole populations depends on whether its efficacy and tolerability can be maintained in further treatment trials.