EMAIL THIS PAGE TO A FRIEND
Applied biochemistry and biotechnology
Single-step conversion of cephalosporin-C to 7-aminocephalosporanic acid by free and immobilized cells of Pseudomonas diminuta.
PMID 16014995
Abstract
7-Aminocephalosporanic acid (7-ACA), the starting material for the production of a number of clinically used semisynthetic cephalosporins, is produced by deacylation of cephalosporin-C. The production of 7-ACA was studied in various modes, at the optimal conditions using free and immobilized whole cells of Pseudomonas diminuta.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
191140
7-Aminocephalosporanic acid, 98%
C10H12N2O5S