EMAIL THIS PAGE TO A FRIEND
The Journal of antimicrobial chemotherapy
Toxicity of cefpirome: an overview.
PMID 1601761
Abstract
Cefpirome is a new cephalosporin antibiotic with a broad antimicrobial spectrum in vitro. This includes strains which are frequently resistant to other cephalosporins (Seibert et al., 1983). This report gives a summary of the toxicological investigations on cefpirome.