EMAIL THIS PAGE TO A FRIEND
The Canadian veterinary journal = La revue veterinaire canadienne
A degenerative encephalomyelopathy in 7 Kuvasz puppies.
PMID 16018562
Abstract
Seven Kuvasz puppies from 2 same-parentage litters developed weakness and ataxia. Six necropsied dogs had lesions in caudate nucleus, cerebellar nuclei and folia, and spinal cord. Lesions seen were felt to be familial or due to the effects of an amprolium-induced thiamine deficiency on the developing brains of these puppies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0542
Amprolium hydrochloride
C14H19ClN4 · HCl
31592
Amprolium hydrochloride, VETRANAL, analytical standard
C14H19ClN4 · HCl