EMAIL THIS PAGE TO A FRIEND
Farmaco (Societa chimica italiana : 1989)
High performance liquid chromatographic determination of oxeladin citrate and oxybutynin hydrochloride and their degradation products.
PMID 16023117
Abstract
Two high performance liquid chromatographic (HPLC) methods are presented for the determination of oxeladin citrate (OL) and oxybutynin hydrochloride (OB) and their degradation products. The first method was based on HPLC separation of OL from its degradation product using a Nucleosil C(18) column with a mobile phase consisting of acetonitrile -0.1% phosphoric acid (60:40 v/v). The second method was based on HPLC separation of OB from its degradation product using a VP-ODS C(18) column with a mobile phase consisting of acetonitrile/0.01 M potassium dihydrogen phosphate/diethylamine (60:40:0.2). Quantitation was achieved with UV detection at 220 nm based on peak area. The two HPLC methods were applied for the determination of OL or OB, their degradation products, methylparaben and propylparaben in pharmaceutical preparations. The proposed methods were used to investigate the kinetics of acidic and alkaline degradation processes of OL and OB at different temperatures and the apparent pseudofirst-order rate constant, half-life and activation energy were calculated. The pH-rate profiles of degradation of OL and OB in Britton-Robinson buffer solutions within the pH range 2-12 were studied.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O3878
Oxeladin citrate salt, analytical standard
C20H33NO3 · C6H8O7