EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Ku86 preserves chromatin integrity in cells adapted to high NaCl.
PMID 16027367
Abstract
Cells adapted to high NaCl have many DNA breaks both in cell culture and in the renal inner medulla in vivo; yet they survive, function, and even proliferate. Here, we show that Ku86 is important for maintaining chromosomal integrity despite the continued presence of DNA breaks. The Ku heterodimer is part of DNA-dependent PK (DNA-PK), a complex that contributes by nonhomologous end joining to repair of double-strand breaks. We demonstrate that cells deficient in Ku86, but not cells deficient in DNA-PKcs (the catalytic subunit of DNA-PK), are hypersensitive to high NaCl as manifested by profound inhibition of proliferation, aberrant mitosis, and increased chromosomal fragmentation. Lower eukaryotes, including the soil nematode Caenorhabditis elegans, lack a DNA-PKcs homologue but are able to adapt to high NaCl. We show that cells of C. elegans adapted to high NaCl have many DNA breaks, similar to the mammalian cells adapted to high NaCl. Ku86 mutant C. elegans as well as C. elegans fed with cku86 dsRNA also display hypersensitivity to high NaCl, characterized by a reduced number of progeny and prolonged generation time in high NaCl. We propose that Ku86 ameliorates the effects of high NaCl-induced DNA breaks in adapted cells by supporting alignment of the broken ends of the DNA and thus maintaining integrity of the fragmented chromatin.