EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Pseudouridine detection improvement by derivatization with methyl vinyl sulfone and capillary HPLC-mass spectrometry.
PMID 16029966
Abstract
A method is presented for improved detection of pseudouridine in nucleoside mixtures based on the specific derivatization with methyl vinyl sulfone followed by analysis by capillary HPLC-mass spectrometry. Reaction conditions were optimized in order to obtain the best yield and specificity. The method was successfully applied to different nucleoside mixtures.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
247197
Methyl vinyl sulfone, ≥97%
C3H6O2S