EMAIL THIS PAGE TO A FRIEND
Orthopaedic review
Oxaprozin: a new NSAID.
PMID 1603605
Abstract
Oxaprozin, a propionic acid derivative, is a non-steroidal anti-inflammatory drug (NSAID) with analgesic and antipyretic properties. Its spectrum of anti-inflammatory activity has been demonstrated in both in vitro and in vivo models.1 The majority of published clinical trials have focused on the use of oxaprozin (from Searle) in the treatment of osteoarthritis and rheumatoid arthritis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O9637
Oxaprozin, solid
C18H15NO3
1482207
Oxaprozin, United States Pharmacopeia (USP) Reference Standard
C18H15NO3