EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Conformation-dependent inactivation of human betaine-homocysteine S-methyltransferase by hydrogen peroxide in vitro.
PMID 16038618
Abstract
Betaine-homocysteine S-methyltransferase (BHMT) transfers a methyl group from betaine to Hcy to form DMG (dimethylglycine) and Met. The reaction is ordered Bi Bi; Hcy is the first substrate to bind and Met is the last product off. Using intrinsic tryptophan fluorescence [Castro, Gratson, Evans, Jiracek, Collinsova, Ludwig and Garrow (2004) Biochemistry 43, 5341-5351], it was shown that BHMT exists in three steady-state conformations: enzyme alone, enzyme plus occupancy at the first substrate-binding site (Hcy or Met), or enzyme plus occupancy at both substrate-binding sites (Hcy plus betaine, or Hcy plus DMG). Betaine or DMG alone do not bind to the enzyme, indicating that the conformational change associated with Hcy binding creates the betaine-binding site. CBHcy [S-(d-carboxybutyl)-D,L-homocysteine] is a bisubstrate analogue that causes BHMT to adopt the same conformation as the ternary complexes. We report that BHMT is susceptible to conformation-dependent oxidative inactivation. Two oxidants, MMTS (methyl methanethiosulphonate) and hydrogen peroxide (H2O2), cause a loss of the enzyme's catalytic Zn (Zn2+ ion) and a correlative loss of activity. Addition of 2-mercaptoethanol and exogenous Zn after MMTS treatment restores activity, but oxidation due to H2O2 is irreversible. CD and glutaraldehyde cross-linking indicate that H2O2 treatment causes small perturbations in secondary structure but no change in quaternary structure. Oxidation is attenuated when both binding sites are occupied by CBHcy, but Met alone has no effect. Partial digestion of ligand-free BHMT with trypsin produces two large peptides, excising a seven-residue peptide within loop L2. CBHcy but not Met binding slows down proteolysis by trypsin. These findings suggest that L2 is involved in the conformational change associated with occupancy at the betaine-binding site and that this conformational change and/or occupancy at both ligand-binding sites protect the enzyme from oxidative inactivation.
Related Materials