EMAIL THIS PAGE TO A FRIEND
Steroids
Electrochemical bromination of cholest-5-enes.
PMID 16039678
Abstract
Four 5,6-unsaturated steroids--3beta-chlorocholest-5-ene (1a), cholesterol (1b) and its acetate (1c) and benzoate (1d)-were subjected to constant current electrolysis (50 mA, 2 F mol(-1)) in an electrolytic cell divided by a ceramic membrane, using a platinum foil as the anode and a graphite stick as the cathode. When electrolysis was carried out in a solution of tetraethylammonium bromide in aprotic solvents (dichloromethane, acetonitrile or acetic anhydride), the addition of electrochemically-generated elemental bromine onto the double bond of the cholesterol derivatives gave their corresponding 5alpha,6beta-dibromosteroids--3beta-chloro-5alpha,6beta-dibromocholestane (2a), 5alpha,6beta-dibromocholestan-3beta-ol (2b), 5alpha,6beta-dibromocholestan-3beta-yl acetate (2c) and 5alpha,6beta-dibromocholestan-3beta-yl benzoate (2d)--as the sole products, and in good yields (58-91%). However, the electrolysis of steroids 1a-c in a solution of tetraethylammonium bromide with methanol as the solvent proceeded to give, in addition to dibromides 2a-c, the corresponding diastereomeric pairs of 5-bromo-6-methoxysteroids: 5alpha-bromo-3beta-chloro-6beta-methoxycholestane (3a) and 5beta-bromo-3beta-chloro-6alpha-methoxycholestane (4a), 5alpha-bromo-6beta-methoxycholestan-3beta-ol (3b) and 5beta-bromo-6alpha-methoxycholestan-3beta-ol (4b) and 5alpha-bromo-6beta-methoxycholestan-3beta-yl acetate (3c) and 5alpha-bromo-6beta-methoxycholestan-3beta-yl acetate (4c). The benzoate 1d was not soluble enough in methanol, even with heating. The products were characterized by physical and spectral data (IR, 1H NMR and 13C NMR). Single crystal X-ray structure determinations of compounds 2a and 3a are also reported.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C76604
Cholesteryl chloride, 97%
C27H45Cl
GF41858309
Platinum, foil, 0.2m coil, thickness 0.05mm, coil width 1mm, as rolled, 99.99%
Pt
GF58445433
Platinum, foil, 0.2m coil, thickness 0.125mm, coil width 2.5mm, as rolled, 99.99%
Pt
GF34340408
Platinum, foil, 0.2m coil, thickness 0.15mm, coil width .9mm, as rolled, 99.99%
Pt
GF09598809
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width .4mm, as rolled, 99.95%
Pt
GF07973961
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width .85mm, as rolled, 99.95%
Pt
GF65809222
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width 1.2mm, as rolled, 99.95%
Pt
GF35123307
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width 1.6mm, as rolled, 99.95%
Pt
GF21169526
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width 2mm, as rolled, 99.95%
Pt
GF13420257
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width 5mm, as rolled, 99.95%
Pt
GF60594442
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width 8mm, as rolled, 99.95%
Pt
GF52150801
Platinum, foil, 0.5m coil, thickness 0.029mm, coil width .34mm, annealed, 99.99%
Pt
GF91778024
Platinum, foil, 0.5m coil, thickness 0.030mm, coil width .7mm, as rolled, 99.95%
Pt
GF25102897
Platinum, foil, 0.5m coil, thickness 0.045mm, coil width .75mm, as rolled, 99.95%
Pt
GF04267795
Platinum, foil, 0.5m coil, thickness 0.05mm, coil width 1mm, as rolled, 99.99%
Pt
GF66494795
Platinum, foil, 1.2mm disks, thickness 0.025mm, as rolled, 99.95%
Pt
GF86033167
Platinum, foil, 10mm disks, thickness 0.004mm, as rolled, 99.95%
Pt
GF06751267
Platinum, foil, 10mm disks, thickness 0.005mm, as rolled, 99.95%
Pt
GF96551354
Platinum, foil, 10mm disks, thickness 0.0075mm, as rolled, 99.95%
Pt
GF73706058
Platinum, foil, 10mm disks, thickness 0.0125mm, as rolled, 99.95%
Pt
GF29522101
Platinum, foil, 10mm disks, thickness 0.01mm, as rolled, 99.95%
Pt
GF31105864
Platinum, foil, 10mm disks, thickness 0.025mm, as rolled, 99.95%
Pt
GF06396532
Platinum, foil, 10mm disks, thickness 0.025mm, as rolled, 99.99+%
Pt
GF61547632
Platinum, foil, 8mm disks, thickness 0.01mm, as rolled, 99.95%
Pt
GF11448840
Platinum, foil, 8mm disks, thickness 0.025mm, as rolled, 99.95%
Pt
GF60675442
Platinum, foil, 8mm disks, thickness 0.05mm, as rolled, 99.95%
Pt
GF38543946
Platinum, foil, 8mm disks, thickness 0.125mm, as rolled, 99.95%
Pt
GF65307090
Platinum, foil, 8mm disks, thickness 0.125mm, as rolled, 99.99+%
Pt
GF05324183
Platinum, foil, 8mm disks, thickness 0.1mm, as rolled, 99.99+%
Pt
GF40155533
Platinum, foil, light tested, 25x25mm, thickness 0.005mm, as rolled, 99.95%
Pt