EMAIL THIS PAGE TO A FRIEND
Organic letters
An anionic nucleophilic catalyst system for the diastereoselective synthesis of trans-beta-lactams.
PMID 16048317
Abstract
Trans-disubstituted beta-lactams show increasing utility and prominence in numerous pharmaceutical applications, making their asymmetric synthesis an attractive goal for chemists. We introduce an anionic, nucleophilic catalyst system that provides an efficient, diastereoselective route to trans-disubstituted beta-lactams, a complement to our previously described catalytic methodology for generating the corresponding cis diastereomers. This catalytic, "switch mechanism" process allows for flexibility in the stereoselective synthesis of beta-lactams, producing either cis or trans products as desired from the same substrates. [reaction: see text]
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
462292
N-Benzylethylenediamine, 97%
C9H14N2