EMAIL THIS PAGE TO A FRIEND
Organic letters
Concise synthesis of all stereoisomers of beta-methoxytyrosine and determination of the absolute configuration of the residue in callipeltin A.
PMID 16048348
Abstract
All stereoisomers of beta-methoxytyrosine (beta-OMeTyr), a stereo-undefined component of callipeltin A, were synthesized from L- and D-tyrosine. The stereochemistry of beta-OMeTyr in callipeltin A was determined to be 2R,3R by an oxidative procedure and Marfey's analysis. [structure: see text]
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1420006 Levodopa Related Compound B, United States Pharmacopeia (USP) Reference Standard
C10H13NO4