EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
A Facile method to transform trans-4-carboxy-3,4-dihydro-3-phenyl- 1(2H)-isoquinolones to indeno[1,2-c]isoquinolines.
PMID 16050715
Abstract
The indeno[1,2-c]isoquinolines are an important class of topoisomerase I inhibitors with anticancer activity. The condensation of Schiff bases with homophthalic anhydrides provides a mixture of cis- and trans-4-carboxy-3,4-dihydro-3-phenyl-1(2H)isoquinolones. Although the cis products can be readily converted to indeno[1,2-c]isoquinolines with thionyl chloride, the trans products do not afford indeno[1,2-c]isoquinolines using this method. The present report describes a route for conversion of the trans diastereomers to indeno[1,2-c]isoquinolines using selenoxide elimination and Friedel-Crafts cyclization chemistry.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
293121
Thionyl chloride solution, 2.0 M in methylene chloride
Cl2OS