EMAIL THIS PAGE TO A FRIEND
Cell
Oocyte generation in adult mammalian ovaries by putative germ cells in bone marrow and peripheral blood.
PMID 16051153
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB4301
Anti-Stage-Specific Embryonic Antigen-1 Antibody, clone MC-480