EMAIL THIS PAGE TO A FRIEND
Drug metabolism and disposition: the biological fate of chemicals
Extracts and kavalactones of Piper methysticum G. Forst (kava-kava) inhibit P-glycoprotein in vitro.
PMID 16051732
Abstract
Root extracts from kava-kava (Piper methysticum G. Forst) are clinically used for the treatment of anxiety and restlessness. Due to reported cases of liver toxicity, kava-kava extracts were withdrawn from the market in several countries in 2002. Because the efflux transporter P-glycoprotein (P-gp) is involved in the absorption, distribution, and excretion of many drugs and often participates in drug-drug interactions, we studied the effect of a crude kava extract and the main kavalactones kavain, dihydrokavain, methysticin, dihydromethysticin, yangonin, and desmethoxyyangonin on the P-gp-mediated efflux of calcein-acetoxymethylester in the P-gp-overexpressing cell line P388/dx and the corresponding cell line P388. The crude extract and the kavalactones showed a moderate to potent inhibitory activity with f2) (concentration needed to double baseline fluorescence) values of 170 microg/ml and 17 to 90 microM, respectively. The f2 value of yangonin could not be determined due to its higher lipophilicity. In conclusion, our results for the first time demonstrate P-gp-inhibitory activity of kava-kava and its components in vitro.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
41866
(S)-(+)-7,8-Dihydrokavain, analytical standard
C14H16O3