EMAIL THIS PAGE TO A FRIEND
Compendium of continuing education in dentistry (Jamesburg, N.J. : 1995)
Techniques used to fabricate all-ceramic restorations in the dental practice.
PMID 16060381
Abstract
Porcelain is an increasingly popular material to use for restorations. This article will discuss the 3 main ways to fabricate porcelain restorations. The first method involves waxing up the restoration to the proper form and casting it in molten porcelain similar to the lost wax technique for gold. The second technique requires the use of porcelain in a powder form to be stacked on top of a refractory die or a platinum foil and then fired in the oven. The third main technique is the use of a CAD/CAM system to mill the porcelain restoration from a solid block of porcelain. All 3 techniques are valid and the clinician should have a thorough understanding of which techniques are appropriate in various clinical situations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF41858309
Platinum, foil, 0.2m coil, thickness 0.05mm, coil width 1mm, as rolled, 99.99%
Pt
GF58445433
Platinum, foil, 0.2m coil, thickness 0.125mm, coil width 2.5mm, as rolled, 99.99%
Pt
GF34340408
Platinum, foil, 0.2m coil, thickness 0.15mm, coil width .9mm, as rolled, 99.99%
Pt
GF09598809
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width .4mm, as rolled, 99.95%
Pt
GF07973961
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width .85mm, as rolled, 99.95%
Pt
GF65809222
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width 1.2mm, as rolled, 99.95%
Pt
GF35123307
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width 1.6mm, as rolled, 99.95%
Pt
GF21169526
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width 2mm, as rolled, 99.95%
Pt
GF13420257
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width 5mm, as rolled, 99.95%
Pt
GF60594442
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width 8mm, as rolled, 99.95%
Pt
GF52150801
Platinum, foil, 0.5m coil, thickness 0.029mm, coil width .34mm, annealed, 99.99%
Pt
GF91778024
Platinum, foil, 0.5m coil, thickness 0.030mm, coil width .7mm, as rolled, 99.95%
Pt
GF25102897
Platinum, foil, 0.5m coil, thickness 0.045mm, coil width .75mm, as rolled, 99.95%
Pt
GF04267795
Platinum, foil, 0.5m coil, thickness 0.05mm, coil width 1mm, as rolled, 99.99%
Pt
GF66494795
Platinum, foil, 1.2mm disks, thickness 0.025mm, as rolled, 99.95%
Pt
GF86033167
Platinum, foil, 10mm disks, thickness 0.004mm, as rolled, 99.95%
Pt
GF06751267
Platinum, foil, 10mm disks, thickness 0.005mm, as rolled, 99.95%
Pt
GF96551354
Platinum, foil, 10mm disks, thickness 0.0075mm, as rolled, 99.95%
Pt
GF73706058
Platinum, foil, 10mm disks, thickness 0.0125mm, as rolled, 99.95%
Pt
GF29522101
Platinum, foil, 10mm disks, thickness 0.01mm, as rolled, 99.95%
Pt
GF31105864
Platinum, foil, 10mm disks, thickness 0.025mm, as rolled, 99.95%
Pt
GF06396532
Platinum, foil, 10mm disks, thickness 0.025mm, as rolled, 99.99+%
Pt
GF61547632
Platinum, foil, 8mm disks, thickness 0.01mm, as rolled, 99.95%
Pt
GF11448840
Platinum, foil, 8mm disks, thickness 0.025mm, as rolled, 99.95%
Pt
GF60675442
Platinum, foil, 8mm disks, thickness 0.05mm, as rolled, 99.95%
Pt
GF38543946
Platinum, foil, 8mm disks, thickness 0.125mm, as rolled, 99.95%
Pt
GF65307090
Platinum, foil, 8mm disks, thickness 0.125mm, as rolled, 99.99+%
Pt
GF05324183
Platinum, foil, 8mm disks, thickness 0.1mm, as rolled, 99.99+%
Pt
GF40155533
Platinum, foil, light tested, 25x25mm, thickness 0.005mm, as rolled, 99.95%
Pt
GF02102948
Platinum, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, 99.99+%
Pt