EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Structural basis for bending of organic crystals.
PMID 16075080
Abstract
Bending is observed in organic crystals when the packing is anisotropic in such a way that strong and weak interaction patterns occur in nearly perpendicular directions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
251089
2-(Methylthio)pyridine-3-carboxylic acid, 98%
C7H7NO2S