EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Highly enantioselective catalytic thiolysis of prochiral cyclic dicarboxylic anhydrides utilizing a bifunctional chiral sulfonamide.
PMID 16080228
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
519367
3,5-Bis(trifluoromethyl)benzenesulfonyl chloride, 97%
C8H3ClF6O2S