EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
In situ formation of ligand and catalyst--application in ruthenium-catalyzed enantioselective reduction of ketones.
PMID 16091792
Abstract
The direct in situ formation of highly efficient ruthenium-catalysts for the asymmetric reduction of ketones was obtained by combining chiral ligand building blocks with a ruthenium precursor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
684414
Boc-L-alanine (2S)-2-hydroxylpropylamide, 97%
C11H22N2O4