EMAIL THIS PAGE TO A FRIEND
International journal of cancer
Methylation of the O6-methylguanine-DNA methyltransferase promoter suppresses expression in mouse skin tumors and varies with the tumor induction protocol.
PMID 16094607
Abstract
Hypermethylation of CpG sites within the promoter region of the O6-methylguanine-DNA methyltransferase (MGMT) gene occurs frequently in human cancer, preventing both MGMT expression and repair of alkylation damage. To assess the role of MGMT in the development of mouse skin tumors induced by initiation-promotion protocols, methylation of the MGMT promoter was examined in tumor DNA using methylation-specific PCR. To determine whether MGMT promoter methylation was affected by the tumor induction protocol, tumors were initiated by N-methyl-N'-nitro-N-nitrosoguanidine (MNNG) or 7,12-dimethylbenz[a]anthracene (DMBA) and promoted by 12-O-tetradecanoylphorbol-13-acetate (TPA) or mezerein. Although the MGMT promoter was not methylated in normal skin, promoter methylation was found in 56 of 136 papillomas (41.2%) and in 19 of 37 squamous cell carcinomas (51.4%). When methylation of the MGMT promoter was compared in the 4 treatment groups, hypermethylation was found more frequently in tumors initiated by DMBA and promoted by mezerein, a protocol associated with a high frequency of malignant conversion. Methylation was found in some tumors as early as 5 weeks after initiation, but the methylation frequency increased with time. MGMT promoter methylation reduced MGMT expression as determined by immunohistochemistry. Although MGMT promoter methylation was not generally correlated with ras mutations, the frequency of MGMT methylation was higher in MNNG-initiated, mezerein-promoted papillomas with mutations in Ha-ras compared to papillomas with Ki-ras. Methylation of the MGMT promoter, associated with reduced MGMT expression, is found in nearly half of mouse skin tumors, but varies with both the tumor initiator and tumor promoter, and may be a key step in the progression from papillomas to carcinomas.