EMAIL THIS PAGE TO A FRIEND
FEMS microbiology letters
Toxicity testing of destruxins and crude extracts from the insect-pathogenic fungus Metarhizium anisopliae.
PMID 16102914
Abstract
Increasing sensitivity towards secondary metabolites from fungal biological control agents (BCAs) has prompted the toxicological risk assessment of metabolites produced by the insect pathogenic fungus Metarhizium anisopliae. Viability studies on one human and one insect cell line were used to compare the two approaches of testing individual metabolites (destruxins A, B and E) or the complete crude extract from liquid cultures. Furthermore, crude extract was separated into fractions, which did not contain the main destruxins A, B and E. Evaluation of the cytotoxic activity of these different compounds suggested that a wide range of metabolites with synergistic or adverse effects are present in the crude extract. The results indicate that identification and toxicological assessment of each individual metabolite produced by a BCA is not only time and cost-intensive, but also does not convey the whole picture. Testing of the crude extract offers an alternative approach and is recommended when assessing the risks of metabolites for registration purposes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D4921
Destruxin A from Metarrhizium anisopliae, ≥98% (HPLC)
C29H47N5O7