EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
A new reactivity pattern for vinyl bromides: cine-substitution via palladium catalysed C-N coupling/Michael addition reactions.
PMID 16106288
Abstract
Palladium catalysed C-N bond formation can be used to convert vinyl bromides to the corresponding enamines, which are reacted in situ with alkylidene malonates to provide Michael adducts. The overall transformation results in cine-substitution of the starting vinyl bromide.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V1902
Vinyl bromide, 98%
C2H3Br
434191
Vinyl bromide solution, 1.0 M in THF
C2H3Br