EMAIL THIS PAGE TO A FRIEND
Journal of environmental monitoring : JEM
Organophosphate triesters in indoor environments.
PMID 16121268
Abstract
In this study the occurrence of ten organophosphate triesters in indoor air at 29 different locations was investigated. They were detected at all locations and a total of ten compounds were identified. The predominant compounds were the chlorinated compounds tris(chloroisopropyl) phosphate and tris(2-chloroethyl) phosphate with a concentration around 2 microg m(-3) in some indoor environments. A rough estimation gives at hand a daily inhalation of around 10 microg for each of these two compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
119660
Tris(2-chloroethyl) phosphate, 97%
C6H12Cl3O4P