EMAIL THIS PAGE TO A FRIEND
Drug metabolism and disposition: the biological fate of chemicals
Gefitinib (Iressa) inhibits the CYP3A4-mediated formation of 7-ethyl-10-(4-amino-1-piperidino)carbonyloxycamptothecin but activates that of 7-ethyl-10-[4-N-(5-aminopentanoic acid)-1-piperidino]carbonyloxycamptothecin from irinotecan.
PMID 16123050
Abstract
Gefitinib (Iressa) is an anticancer drug that selectively inhibits tyrosine kinases of epidermal growth factor receptor. Gefitinib might affect CYP3A4-mediated metabolism, since the drug is a substrate of human CYP3A. In this study, we evaluated the effects of gefitinib on drug metabolism catalyzed by human CYP3A4. The effects of gefitinib on the CYP3A4-mediated formation of NPC (7-ethyl-10-(4-amino-1-piperidino)carbonyloxycamptothecin) and that of APC (7-ethyl-10-[4-N-(5-aminopentanoic acid)-1-piperidino]carbonyloxycamptothecin) from irinotecan were examined with the use of human liver and small intestinal microsomes. Gefitinib inhibited the formation of NPC in liver and small intestinal microsomes. The apparent intrinsic metabolic clearance (CL(int)) in the presence of 40 microM gefitinib was equivalent to about 26% of control in liver microsomes and 45% of control in small intestinal microsomes. Gefitinib stimulated the formation of APC by CYP3A4. CL(int) in the presence of 20 microM gefitinib with human liver microsomes was about 1.9 times higher than control. In human small intestinal microsomes, APC formation was enhanced by the addition of gefitinib at concentrations 20 microM or higher. CL(int) in the presence of 40 microM gefitinib was 2.8 times higher than control. Thus, we discovered that gefitinib inhibited the formation of NPC but stimulated the formation of APC from irinotecan.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
123188
5-Aminovaleric acid, 97%
C5H11NO2