EMAIL THIS PAGE TO A FRIEND
Journal of chemical ecology
Do naïve ruminants degrade alkaloids in the rumen?
PMID 16124250
Abstract
Three different methods for the culture of rumen microorganisms (Hungate's technique, the Hohenheim in vitro gas production method, and the semicontinuous rumen simulation technique) were employed to study the influence of various alkaloids (sparteine, lupanine, cytisine, atropine, quinidine, lobeline, harmaline, arecoline, nicotine, caffeine, pilocarpine, gramine, senecionine, and monocrotaline) on rumen microorganisms. Rumen micro-organisms from naive ruminants (sheep, cattle) that had not been exposed to the alkaloids before were generally not able to degrade most of the alkaloids. Only the alkaloids pilocarpine, gramine, and monocrotaline appeared to be degradable. Rumen microorganisms from a sheep preconditioned to lupin alkaloids tolerated lupanine much better than nonadapted microorganisms, but no degradation occurred. The findings indicate that the main site of detoxification in naive ruminants is not the rumen but more likely the liver and kidneys as in nonruminants.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G10806
Gramine, 99%
C11H14N2