EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Microbial transformation of curvularin.
PMID 16124776
Abstract
The microbiological transformation of curvularin (1) with Beauveria bassiana ATCC 7159 afforded three new metabolites identified as curvularin-7-O-beta-D-glucopyranoside (2), curvularin-4'-O-methyl-7-O-beta-D-glucopyranoside (3), and 6-hydroxycurvularin-4'-O-methyl-6-O-beta-D-glucopyranoside (4) resulting from hydroxylation, glucosidation, and methylglucosidation of the substrate. Isolation of 6-methyl-2,4-dihydroxyphenyl-4'-O-methyl-1-O-beta-D-glucopyranoside (5) from the fermentation broth of B. bassiana ATCC 7159 without any added substrate suggested that hydroxylase, glucosyl transferase, and methylase are constitutive enzymes of this organism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0772
Curvularin, from Penicillium citrinum, ≥98% (HPLC)
C16H20O5