EMAIL THIS PAGE TO A FRIEND
International urology and nephrology
Selective embolization in the management of arteriovenous fistula after renal allograft biopsy preserves renal allograft function.
PMID 16132785