EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Joys of molecules. 2. Endeavors in chemical biology and medicinal chemistry.
PMID 16134928
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E3656
(−)-Epothilone A, from Sorangium cellulosum, >95% (HPLC), solid
C26H39NO6S