EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Urea- and thiourea-substituted cinchona alkaloid derivatives as highly efficient bifunctional organocatalysts for the asymmetric addition of malonate to nitroalkenes: inversion of configuration at C9 dramatically improves catalyst performance.
PMID 16136619