EMAIL THIS PAGE TO A FRIEND
Neuropediatrics
Possible genotype-phenotype correlations in children with mild clinical course of Canavan disease.
PMID 16138249
Abstract
Canavan disease is characterised as a rare, neurodegenerative disease that usually causes death in early childhood. It is an autosomal recessive disorder due to an aspartoacylase (ASPA) deficiency. The causative gene has been mapped to chromosome 17 pter-p13. Here we describe three affected children from two Greek families with an unusually mild course of Canavan disease. All children presented with muscular hypotonia and macrocephaly. Diagnosis was based on elevated N-acetylaspartate in urine, reduced aspartoacylase activity in fibroblasts, and marked white matter changes on cerebral imaging. All three affected individuals exhibited continuous psychomotor development without any regression. Genetic analyses revealed compound heterozygous mutations (Y288 C; F295 S) in two individuals. The Y288 C variant was previously described in a child with macrocephaly, mild developmental delay, increased signal intensity in the basal ganglia, partial cortical blindness and retinitis pigmentosa, and slightly elevated N-acetylaspartate in the urine. Demonstration of the same variant in two unusually mildly affected Canavan disease patients and absence of this variant in 154 control chromosomes suggest a possible pathogenic role in mild Canavan disease. In the third individual, two homozygous sequence variants were identified, which comprise the known G274R mutation and a novel K213E variant.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00920
N-Acetyl-L-aspartic acid, ≥99.0% (T)
C6H9NO5