EMAIL THIS PAGE TO A FRIEND
Journal of magnetic resonance (San Diego, Calif. : 1997)
Improving pulse sequences for 3D DOSY: convection compensation.
PMID 16140552
Abstract
Signal overlap in the NMR dimension significantly complicates the construction and analysis of 2D diffusion-ordered (DOSY) spectra. Such problems can often be reduced or even eliminated by extending the NMR domain of a DOSY experiment into two dimensions, giving a 3D-DOSY spectrum. To date such experiments have generally sacrificed some signal-to-noise ratio and have required extensive and time-consuming phase cycling. A new family of pulse sequences with internal diffusion encoding (IDOSY) has been introduced which avoids both of these problems. It is often straightforward to incorporate convection compensation in such sequences at no cost in signal-to-noise ratio. Here, some of the problems caused by convection in DOSY are described and illustrated, and the efficacy of convection compensation in the 2DJ-IDOSY and COSY-IDOSY experiments is demonstrated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
456055
Camphene, 95%
C10H16
442505
Camphene, analytical standard
C10H16